Trihexyphenidyl hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

T3303-25G T3303-5G
TCT3303-25GEA 526.14 CAD
TCT3303-25G TCT3303-5G
Trihexyphenidyl hydrochloride ≥98.0% (by HPLC, titration analysis)
Trihexyphenidyl hydrochloride
Formula: C₂₀H₃₁NO·HCl
MDL Number: MFCD00058212
CAS Number: 52-49-3
Merck Index: 14,09695

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR