1-(2-Propynyl)piperazine ≥97.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
1-(Prop-2-yn-1-yl)piperazine, 1-Propargylpiperazine
P2228-1G
P2228-200MG
76220-218EA
588.09
CAD
76220-218
76220-220
1-(2-Propynyl)piperazine ≥97.0% (by GC, titration analysis)
1-(2-Propynyl)piperazine
Formula:
C₇H₁₂N₂ Boiling Pt: 90°C |
MDL Number:
MFCD08445213 CAS Number: 52070-67-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...