[3-(6-Aminohexylamino)propyl]trimethoxysilane ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A3128-25ML A3128-5ML
76219-298EA 733.12 CAD
76219-298 76219-300
[3-(6-Aminohexylamino)propyl]trimethoxysilane ≥97.0% (by GC, titration analysis)
[3-(6-Aminohexylamino)propyl]trimethoxysilane
Formula: C₁₂H₃₀N₂O₃Si
MDL Number: MFCD00054943
CAS Number: 51895-58-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR