Ethyl Isopropyl sulfide ≥98.0% (by GC)

Supplier: TCI America
Danger

E0225-25G E0225-5G
TCE0225-25GEA 262.28 CAD
TCE0225-25G TCE0225-5G
Ethyl Isopropyl sulfide ≥98.0% (by GC)
Ethyl Isopropyl sulfide
Formula: C₅H₁₂S
MW: 104.21 g/mol
Boiling Pt: 103°C
MDL Number: MFCD00039908
CAS Number: 5145-99-3
EINECS: 225-917-5
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR