2-Bromo-5-decylthiophene ≥97.0% (by GC)
Supplier: TCI America
B5406-1G
B5406-5G
TCB5406-1GEA
418.7
CAD
TCB5406-1G
TCB5406-5G
2-Bromo-5-decylthiophene ≥97.0% (by GC)
2-Bromo-5-decylthiophene
Formula:
C₁₄H₂₃BrS Boiling Pt: 135°C |
CAS Number:
514188-72-8 |
Specification Test Results
Appearance | White to Almost white clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...