Undecylphosphonic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

U0088-1G U0088-5G
TCU0088-1GEA 342.61 CAD
TCU0088-1G TCU0088-5G
Undecylphosphonic acid ≥98.0% (by titrimetric analysis)
Undecylphosphonic acid
Formula: C₁₁H₂₅O₃P
MDL Number: MFCD00015831
CAS Number: 5137-69-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 100.0 to 104.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR