Tris Dodecyl sulfate ≥96.0% (by titrimetric analysis) for biochemical research

Supplier: TCI America

T3071-1G T3071-250MG
TCT3071-1GEA 541.94 CAD
TCT3071-1G TCT3071-250MG
Tris Dodecyl sulfate ≥96.0% (by titrimetric analysis) for biochemical research
Tris Dodecyl sulfate
Formula: C₁₂H₂₆O₄S·C₄H₁₁NO₃
MDL Number: MFCD00043277
CAS Number: 50996-85-5
EINECS: 256-903-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization back titration) min. 96.0 %
Average molecular weight 384 to 390
Suitability for protein-analysis to pass test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR