1,3,5-Tri(p-tolyl)benzene ≥96.0% (by GC)

Supplier: TCI America

T3370-1G T3370-5G
76220-588EA 180.12 CAD
76220-588 76220-590
1,3,5-Tri(p-tolyl)benzene ≥96.0% (by GC)
1,3,5-Tri(p-tolyl)benzene
Formula: C₂₇H₂₄
MDL Number: MFCD00192798
CAS Number: 50446-43-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 175.0 to 179.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR