nitro-Grela ≥97.0% (by HPLC)

Supplier: TCI America

N1060-100MG N1060-500MG
TCN1060-100MGEA 347.17 CAD
TCN1060-100MG TCN1060-500MG
nitro-Grela ≥97.0% (by HPLC)
nitro-Grela
Formula: C₃₁H₃₇Cl₂N₃O₃Ru
MDL Number: MFCD28411645
CAS Number: 502964-52-5

Order Now

Specification Test Results

Appearance Green to Dark green powder to crystal
Purity (HPLC) min. 97.0 area%
Elemental analysis (Nitrogen) 5.90 to 6.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR