Bis(9,9-dimethyl-9H-fluoren-2-yl)amine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B4664-1G B4664-5G
TCB4664-1GEA 218.04 CAD
TCB4664-1G TCB4664-5G
Bis(9,9-dimethyl-9H-fluoren-2-yl)amine ≥98.0% (by HPLC, total nitrogen)
Bis(9,9-dimethyl-9H-fluoren-2-yl)amine
Formula: C₃₀H₂₇N
MDL Number: MFCD23135317
CAS Number: 500717-23-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 182.0 to 186.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR