1-(4-Propylphenyl)-2,5-di(2-thienyl)-1H-pyrrole ≥98.0% (by GC)

Supplier: TCI America

P2210-1G P2210-200MG
76220-214EA 1071.24 CAD
76220-214 76220-216
1-(4-Propylphenyl)-2,5-di(2-thienyl)-1H-pyrrole ≥98.0% (by GC)
1-(4-Propylphenyl)-2,5-di(2-thienyl)-1H-pyrrole
Formula: C₂₁H₁₉NS₂
CAS Number: 499793-84-9

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 145.0 to 149.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR