Ethyl 3-(4-Cyanophenyl)-3-oxopropionate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Ethyl 3-(4-cyanophenyl)-3-oxopropanoate, 3-(4-Cyanophenyl)-3-oxopropionic acid Ethyl ester

E1080-1G E1080-200MG
TCE1080-1GEA 790.16 CAD
TCE1080-1G TCE1080-200MG
Ethyl 3-(4-Cyanophenyl)-3-oxopropionate ≥98.0% (by GC, titration analysis)
Ethyl 3-(4-Cyanophenyl)-3-oxopropionate
Formula: C₁₂H₁₁NO₃
MW: 217.22 g/mol
Boiling Pt: 140°C
MDL Number: MFCD00029621
CAS Number: 49744-93-6

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR