2-(2-Nitroanilino)ethanol ≥98.0% (by GC)

Supplier: TCI America
Warning

N1000-25G N1000-5G
TCN1000-25GEA 441.29 CAD
TCN1000-25G TCN1000-5G
2-(2-Nitroanilino)ethanol ≥98.0% (by GC)
2-(2-Nitroanilino)ethanol
Formula: C₈H₁₀N₂O₃
MDL Number: MFCD00239475
CAS Number: 4926-55-0
EINECS: 225-555-8

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR