Propylamine hydrobromide ≥97.0% (by total nitrogen and titration analysis)

Supplier: TCI America
Warning

P2502-1G P2502-5G
TCP2502-1GEA 44.24 CAD
TCP2502-1G TCP2502-5G
Propylamine hydrobromide ≥97.0% (by total nitrogen and titration analysis)
Propylamine hydrobromide
Formula: C₃H₉N·HBr
Melting Pt: 182°C
MDL Number: MFCD30749306
CAS Number: 4905-83-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 97.0 %
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR