2-[2-(2-Methoxyethoxy)ethoxy]ethyl acrylate ≥95.0% (by GC) stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M3094-100G M3094-500G
TCM3094-100GEA 169.06 CAD
TCM3094-100G TCM3094-500G
2-[2-(2-Methoxyethoxy)ethoxy]ethyl acrylate ≥95.0% (by GC) stabilized
2-[2-(2-Methoxyethoxy)ethoxy]ethyl acrylate
Formula: C₁₀H₁₈O₅
MDL Number: MFCD30470140
CAS Number: 48067-72-7
EINECS: 256-348-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %
Acid Value max. 2.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR