Di-tert-butyl(methyl)phosphonium tetrafluoroborate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: (t-Bu)2PMeHBF4, Ditert-butyl(methyl)phosphonium tetrafluoroborate, Bis(1,1-dimethylethyl)methylphosphine tetrafluoroborate

D4731-1G D4731-5G
TCD4731-1GEA 216.46 CAD
TCD4731-1G TCD4731-5G
Di-tert-butyl(methyl)phosphonium tetrafluoroborate ≥98.0% (by titrimetric analysis)
Di-tert-butyl(methyl)phosphonium tetrafluoroborate
Formula: C₉H₂₂BF₄P
MW: 248.05 g/mol
MDL Number: MFCD03840579
CAS Number: 479094-62-7
UN: 1759
ADR: 8 III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Iodometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR