9,10-Bis[N-(2-naphthyl)anilino]anthracene ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
B5023-1G
B5023-200MG
TCB5023-1GEA
527.72
CAD
TCB5023-1G
TCB5023-200MG
9,10-Bis[N-(2-naphthyl)anilino]anthracene ≥98.0% (by HPLC, total nitrogen)
9,10-Bis[N-(2-naphthyl)anilino]anthracene
Formula:
C₄₆H₃₂N₂ |
MDL Number:
MFCD12022490 CAS Number: 473717-08-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...