1-Isochromanone ≥98.0% (by GC)

Supplier: TCI America
Warning

I0953-1G I0953-5G
TCI0953-1GEA 97.96 CAD
TCI0953-1G TCI0953-5G
1-Isochromanone ≥98.0% (by GC)
1-Isochromanone
Formula: C₉H₈O₂
Boiling Pt: 160°C
MDL Number: MFCD00799232
CAS Number: 4702-34-5
EINECS: 225-179-4

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR