Triptycene-9,10-dicarboxaldehyde ≥96.0% (by GC)
Supplier: TCI America
T3146-1G
T3146-200MG
TCT3146-1GEA
763.87
CAD
TCT3146-1G
TCT3146-200MG
Triptycene-9,10-dicarboxaldehyde ≥96.0% (by GC)
Triptycene-9,10-dicarboxaldehyde
Formula:
C₂₂H₁₄O₂ |
CAS Number:
467429-73-8 |
Specification Test Results
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...