Hederagenin ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3beta,4alpha-3,23-Dihydroxyolean-12-en-28-oic acid

H1645-200MG H1645-50MG
TCH1645-200MGEA 813.7 CAD
TCH1645-200MG TCH1645-50MG
Hederagenin ≥98.0% (by HPLC, titration analysis)
Hederagenin
Formula: C₃₀H₄₈O₄
Melting Pt: 334°C
Storage Temperature: Refrigerator
MDL Number: MFCD00017385
CAS Number: 465-99-6
EINECS: 207-369-9
Merck Index: 14,04624

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +81.0 to +85.0 deg (C=1, Pyridine)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR