1-tert-Butylimidazole ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4985-25G B4985-5G
76219-480EA 1301.92 CAD
76219-480 76219-482
1-tert-Butylimidazole ≥98.0% (by GC, titration analysis)
1-tert-Butylimidazole
Formula: C₇H₁₂N₂
Boiling Pt: 105°C
MDL Number: MFCD19441219
CAS Number: 45676-04-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR