Aniline Hydroiodide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

A2778-1G A2778-5G
TCA2778-1GEA 120.08 CAD
TCA2778-1G TCA2778-5G
Aniline Hydroiodide ≥98.0% (by HPLC, total nitrogen)
Aniline Hydroiodide
Formula: C₆H₇N·HI
CAS Number: 45497-73-2
EINECS: 256-239-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR