4,4-Diphenyl-2-cyclohexen-1-one ≥98.0% (by GC)

Supplier: TCI America

D4982-1G D4982-5G
TCD4982-1GEA 199.08 CAD
TCD4982-1G TCD4982-5G
4,4-Diphenyl-2-cyclohexen-1-one ≥98.0% (by GC)
4,4-Diphenyl-2-cyclohexen-1-one
Formula: C₁₈H₁₆O
MDL Number: MFCD00075159
CAS Number: 4528-64-7

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 94.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR