N-(4-Biphenylyl)-1-naphthylamine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: N-([1,1'-Biphenyl]-4-yl)naphthalen-1-amine

B5047-1G B5047-200MG
TCB5047-1GEA 418.7 CAD
TCB5047-1G TCB5047-200MG
N-(4-Biphenylyl)-1-naphthylamine ≥98.0% (by GC)
N-(4-Biphenylyl)-1-naphthylamine
Formula: C₂₂H₁₇N
MW: 295.39 g/mol
Boiling Pt: 315°C
MDL Number: MFCD28138082
CAS Number: 446242-37-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR