Pigment Orange 43 ≥94.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: Benzo[lmn]benzo[4,5]imidazo[2,1-b]benzo[4,5]imidazo[2,1-i][3,8]phenanthroline-8,17-dione, Vat Orange 7

P2124-25G P2124-5G
TCP2124-25GEA 436.4 CAD
TCP2124-25G TCP2124-5G
Pigment Orange 43 ≥94.0% (by total nitrogen basis)
Pigment Orange 43
Formula: C₂₆H₁₂N₄O₂
MW: 412.41 g/mol
MDL Number: MFCD00071829
CAS Number: 4424-06-0
EINECS: 224-597-4

Order Now

Specification Test Results

Appearance Orange to Brown powder to crystal
Purity (with Total Nitrogen) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR