4,4'-Dibromo-2-nitrobiphenyl ≥98.0% (by GC)

Supplier: TCI America
Warning

D5189-1G D5189-5G
TCD5189-1GEA 127.98 CAD
TCD5189-1G TCD5189-5G
4,4'-Dibromo-2-nitrobiphenyl ≥98.0% (by GC)
4,4'-Dibromo-2-nitrobiphenyl
Formula: C₁₂H₇Br₂NO₂
MDL Number: MFCD05854318
CAS Number: 439797-69-0

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR