4-(3-Bromophenyl)pyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

B4616-1G B4616-5G
TCB4616-1GEA 156.42 CAD
TCB4616-1G TCB4616-5G
4-(3-Bromophenyl)pyridine ≥98.0% (by GC)
4-(3-Bromophenyl)pyridine
Formula: C₁₁H₈BrN
MDL Number: MFCD16250222
CAS Number: 4373-72-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR