Bevantolol hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America
Warning

B5372-1G B5372-250MG
TCB5372-1GEA 267.02 CAD
TCB5372-1G TCB5372-250MG
Bevantolol hydrochloride ≥98.0% (by HPLC)
Bevantolol hydrochloride
Formula: C₂₀H₂₇NO₄·HCl
MDL Number: MFCD00941389
CAS Number: 42864-78-8
Merck Index: 14,01192

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %
Melting point 139.0 to 144.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR