Phenyl trifluoromethyl sulfone ≥98.0% (by GC)

Supplier: TCI America
Warning

P2195-1G P2195-5G
TCP2195-1GEA 109.02 CAD
TCP2195-1G TCP2195-5G
Phenyl trifluoromethyl sulfone ≥98.0% (by GC)
Phenyl trifluoromethyl sulfone
Formula: C₇H₅F₃O₂S
Boiling Pt: 203°C
MDL Number: MFCD00159083
CAS Number: 426-58-4

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR