2,7-Diphenyl-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

D5427-1G D5427-5G
76219-968EA 208.56 CAD
76219-968 76219-970
2,7-Diphenyl-9H-carbazole ≥98.0% (by GC)
2,7-Diphenyl-9H-carbazole
Formula: C₂₄H₁₇N
MDL Number: MFCD30478534
CAS Number: 42448-04-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR