Methyl 3-Hydroxyphenylacetate ≥98.0% (by GC)

Supplier: TCI America
Warning

M2925-1G M2925-5G
TCM2925-1GEA 83.74 CAD
TCM2925-1G TCM2925-5G
Methyl 3-Hydroxyphenylacetate ≥98.0% (by GC)
Methyl 3-Hydroxyphenylacetate
Formula: C₉H₁₀O₃
Boiling Pt: 146°C
MDL Number: MFCD00017660
CAS Number: 42058-59-3

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR