6-Bromo-4-hydroxycoumarin ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B4699-1G B4699-5G
TCB4699-1GEA 69.52 CAD
TCB4699-1G TCB4699-5G
6-Bromo-4-hydroxycoumarin ≥98.0% (by GC, titration analysis)
6-Bromo-4-hydroxycoumarin
Formula: C₉H₅BrO₃
Melting Pt: 277°C
MDL Number: MFCD00239372
CAS Number: 4139-61-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR