3-(Acetylthio)propionic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A3134-1G A3134-5G
TCA3134-1GEA 135.88 CAD
TCA3134-1G TCA3134-5G
3-(Acetylthio)propionic acid ≥98.0% (by GC, titration analysis)
3-(Acetylthio)propionic acid
Formula: C₅H₈O₃S
Boiling Pt: 128°C
MDL Number: MFCD06208418
CAS Number: 41345-70-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 52.0 to 56.0 deg-C
Solubility in Methanol very faint turbidity

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR