4-Methyl-3-nitrobenzyl alcohol ≥96.0% (by GC)
Supplier: TCI America
M2916-25G
M2916-5G
TCM2916-25GEA
263.86
CAD
TCM2916-25G
TCM2916-5G
4-Methyl-3-nitrobenzyl alcohol ≥96.0% (by GC)
4-Methyl-3-nitrobenzyl alcohol
Formula:
C₈H₉NO₃ Boiling Pt: 195°C |
MDL Number:
MFCD00007177 CAS Number: 40870-59-5 EINECS: 250-120-8 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 41.0 to 45.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...