Ethylene Glycol 1,2-Bis(2-propynyl) ether ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
E1054-25G
E1054-5G
TCE1054-25GEA
1166.04
CAD
TCE1054-25G
TCE1054-5G
Ethylene Glycol 1,2-Bis(2-propynyl) ether ≥97.0% (by GC)
Ethylene Glycol 1,2-Bis(2-propynyl) ether
Formula:
C₈H₁₀O₂ Boiling Pt: 88°C |
MDL Number:
MFCD03640302 CAS Number: 40842-04-4 UN: 3271 ADR: 3,III |
Specification Test Results
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...