PYREX® Reusable Serological Pipettes, Glass, Corning
Supplier: Corning
7085-1X 7085-1 7085-2 7085-2C 7085-10 7085-1C 7085-2X 7085-5
53204-279CS 374.82 CAD
53204-279 53204-315 53204-359 53204-370 53204-417 53204-337 77777-134 89090-934
PYREX® Reusable Serological Pipettes, Glass, Corning
Pipettes Serological Pipettes
Reusable serological pipettes made of PYREX® glass.
  • Calibrated to deliver (TD)
  • Colored graduations are enameled on to the glass
  • Color-coded by size for easy identification and sorting
Tip and top are tempered to provide increased strength. Pipettes are calibrated 'to deliver' and must be blown out to obtain total rated capacity.
Certifications: ASTM E-1044