1,1,2,2,3,3-Hexafluoro-1-(trifluoromethoxy)-3-[(1,2,2-trifluorovinyl)oxy]propane ≥97.0% (by GC)

Supplier: TCI America

H1610-25G H1610-5G
76220-148EA 491.38 CAD
76220-148 76220-150
1,1,2,2,3,3-Hexafluoro-1-(trifluoromethoxy)-3-[(1,2,2-trifluorovinyl)oxy]propane ≥97.0% (by GC)
1,1,2,2,3,3-Hexafluoro-1-(trifluoromethoxy)-3-[(1,2,2-trifluorovinyl)oxy]propane
Formula: C₆F₁₂O₂
Boiling Pt: 64°C
Storage Temperature: Refrigerator
CAS Number: 40573-09-9

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR