1-Bromo-8-iodonaphthalene ≥98.0% (by GC)

Supplier: TCI America
Warning

B5220-1G B5220-200MG
76219-526EA 758.4 CAD
76219-526 76219-528
1-Bromo-8-iodonaphthalene ≥98.0% (by GC)
1-Bromo-8-iodonaphthalene
Formula: C₁₀H₆BrI
Boiling Pt: 168°C
MDL Number: MFCD11656276
CAS Number: 4044-58-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 103.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR