2,8-Dibromodibenzothiophene 5,5-Dioxide ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 2,8-Dibromodibenzothiophene sulfone, 2,8-Dibromodibenzo[b,d]thiophene 5,5-dioxide, 2,8-Dibromodibenzosulfolane, 2,8-Dibromodibenzothiophene sulphone, 2,8-Dibromodibenzosulpholane

D5027-1G D5027-200MG
76219-788EA 507.18 CAD
76219-788 76219-790
2,8-Dibromodibenzothiophene 5,5-Dioxide ≥95.0% (by GC)
2,8-Dibromodibenzothiophene 5,5-Dioxide
Formula: C₁₂H₆Br₂O₂S
MW: 374.05 g/mol
Melting Pt: 361°C
MDL Number: MFCD00093102
CAS Number: 40307-15-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR