9-(2-Biphenylyl)-10-bromoanthracene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 9-([1,1'-Biphenyl]-2-yl)-10-bromoanthracene

B4895-1G B4895-5G
TCB4895-1GEA 231.47 CAD
TCB4895-1G TCB4895-5G
9-(2-Biphenylyl)-10-bromoanthracene ≥98.0% (by GC)
9-(2-Biphenylyl)-10-bromoanthracene
Formula: C₂₆H₁₇Br
MW: 409.33 g/mol
MDL Number: MFCD28138083
CAS Number: 400607-16-1

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 146.0 to 150.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR