Ethyl 2-(4-Hydroxyphenyl)-4-methylthiazole-5-carboxylate hydrochloride ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
E1049-25G
E1049-5G
TCE1049-25GEA
105.86
CAD
TCE1049-25G
TCE1049-5G
Ethyl 2-(4-Hydroxyphenyl)-4-methylthiazole-5-carboxylate hydrochloride ≥98.0% (by HPLC, total nitrogen)
Ethyl 2-(4-Hydroxyphenyl)-4-methylthiazole-5-carboxylate hydrochloride
Formula:
C₁₃H₁₃NO₃S·HCl |
CAS Number:
399017-10-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...