Isoxazolidine hydrochloride ≥97.0% (by titrimetric analysis)

Supplier: TCI America

I0966-1G I0966-5G
TCI0966-1GEA 271.76 CAD
TCI0966-1G TCI0966-5G
Isoxazolidine hydrochloride ≥97.0% (by titrimetric analysis)
Isoxazolidine hydrochloride
Formula: C₃H₇NO·HCl
MDL Number: MFCD11100465
CAS Number: 39657-45-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 125.0 to 129.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR