Carboxymethyldextran sodium salt, M.W. ~ 40000

Supplier: TCI America

C3251-1G C3251-5G
TCC3251-1GEA 64.78 CAD
TCC3251-1G TCC3251-5G
Carboxymethyldextran sodium salt, M.W. ~ 40000
Carboxymethyldextran sodium salt
MDL Number: MFCD00146477
CAS Number: 39422-83-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Content (Sodium) 6.5 to 9.5 % (calcd.on dried substance)
Drying loss max. 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR