2-Fluoro-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

F1182-1G F1182-200MG
TCF1182-1GEA 412.38 CAD
TCF1182-1G TCF1182-200MG
2-Fluoro-9H-carbazole ≥98.0% (by GC)
2-Fluoro-9H-carbazole
Formula: C₁₂H₈FN
MDL Number: MFCD09033508
CAS Number: 391-53-7

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 224.0 to 228.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR