4-(Trifluoromethyl)benzamidine hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America
Warning

T3115-1G T3115-200MG
TCT3115-1GEA 521.4 CAD
TCT3115-1G TCT3115-200MG
4-(Trifluoromethyl)benzamidine hydrochloride ≥98.0% (by HPLC)
4-(Trifluoromethyl)benzamidine hydrochloride
Formula: C₈H₇F₃N₂·HCl
Melting Pt: 167°C
MDL Number: MFCD04114431
CAS Number: 38980-96-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR