2,6-Dibromoanisole ≥98.0% (by GC)

Supplier: TCI America
Warning

D5279-1G D5279-5G
76219-894EA 121.66 CAD
76219-894 76219-896
2,6-Dibromoanisole ≥98.0% (by GC)
2,6-Dibromoanisole
Formula: C₇H₆Br₂O
Boiling Pt: 145°C
Melting Pt: 13°C
MDL Number: MFCD00093270
CAS Number: 38603-09-7

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR