2-(2,4-Dihydroxyphenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

D4819-1G D4819-5G
76219-712EA 159.58 CAD
76219-712 76219-714
2-(2,4-Dihydroxyphenyl)-4,6-diphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
2-(2,4-Dihydroxyphenyl)-4,6-diphenyl-1,3,5-triazine
Formula: C₂₁H₁₅N₃O₂
MW: 341.37 g/mol
Melting Pt: 276°C
MDL Number: MFCD16251126
CAS Number: 38369-95-8

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR