2-Fluoroethyl p-Toluenesulfonate ≥98.0% (by GC)

Supplier: TCI America
Warning

F1056-1G F1056-5G
TCF1056-1GEA 287.56 CAD
TCF1056-1G TCF1056-5G
2-Fluoroethyl p-Toluenesulfonate ≥98.0% (by GC)
2-Fluoroethyl p-Toluenesulfonate
Formula: C₉H₁₁FO₃S
Boiling Pt: 140°C
MDL Number: MFCD01658061
CAS Number: 383-50-6

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR