4-Iodotriphenylamine ≥98.0% (by GC)

Supplier: TCI America
Warning

I0951-1G I0951-5G
TCI0951-1GEA 60.04 CAD
TCI0951-1G TCI0951-5G
4-Iodotriphenylamine ≥98.0% (by GC)
4-Iodotriphenylamine
Formula: C₁₈H₁₄IN
MDL Number: MFCD11046352
CAS Number: 38257-52-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 107.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR