Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
N1029-25G
N1029-5G
TCN1029-25GEA
502.44
CAD
TCN1029-25G
TCN1029-5G
Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid ≥98.0% (by GC, titration analysis)
Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid
Formula:
C₉H₁₀O₄ |
MDL Number:
MFCD00152680 CAS Number: 3813-52-3 EINECS: 223-301-0 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...